Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse Osteoprotegerin/TNFRSF11B Quantikine ELISA Kit
GSA_VASDP

Catalog No. MOP00 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MOP00 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MOP00 Supplier R&D Systems™ Supplier No. MOP00
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Osteoprotegerin/TNFRSF11B Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse Osteoprotegerin/TNFRSF11B Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse Osteoprotegerin/TNFRSF11B in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Assay Range 31.30 to 2000 pg/mL
Assay Sensitivity 6.9 pg/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (EDTA), Serum, Serum
Specificity Natural and Recombinant Mouse OPG
For Use With (Application) ELISA
Gene ID (Entrez) 4982
Gene Symbol Tnfrsf11b
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym OCIF, OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11 B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TNFRSF11B, TR1, tumor necrosis factor receptor superfamily, member 11 b
Target Osteoprotegerin/TNFRSF11B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.