Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse PD-1 His-tag Recombinant Protein

Catalog No. PIRP88247
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP88247 100 μg
PIRP88248 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIRP88247 Supplier Invitrogen™ Supplier No. RP88247
Add to Cart
Add to Cart

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (840 μL) be added to the vial to prepare a stock solution of 1.19 mg/mL. Concentration is measured by UV.

Cell-mediated immune responses are initiated by T lymphocytes that are themselves stimulated by cognate peptides bound to MHC molecules on antig en-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as PD-1 (also known as PDCD-1) that mediate inhibitory signals from the APC. PD-1 can bind two different but related ligands, PDL-1 and PDL-2. Upon binding to either of these ligands, signals generated by PD-1 inhibit the activation of the immune response in the absence of 'danger signals' such as LPS or other molecules associated with bacteria or other pathogens. Evidence for this is seen in PD1-null mice who exhibit hyperactivated immune systems and autoimmune diseases. Despite its predicted molecular weight, PD-1 often migrates at higher molecular weight in SDS-PAGE.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q02242
Concentration 0.25 mg/mL
For Use With (Application) Control, ELISA Standard, Binding Assay
Format Lyophilized
Formulation PBS with 5% mannitol, 5% trehalose and no preservative; pH 7.4
Gene ID (Entrez) 18566
Molecular Weight (g/mol) 40-45 kDa
Name Mouse PD-1 His-tag
pH Range 7.4
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias CD279; EGK_05005; hPD1; hPD-1; hPD-l; hSLE1; Ly101; mPD-1; PD1; PD-1; Pdc1; Pdcd1; programmed cell death 1; programmed cell death 1 protein; programmed cell death protein 1; programmed cell death protein 1-like; programmed death 1; Protein PD1; protein PD-1; sCD279; SLEB2; soluble CD279; systemic lupus erythematosus susceptibility 2
Common Name PD-1 (CD279)
Gene Symbol Pdcd1
Biological Activity Measured by its ability to bind PD-L1 Protein, Mouse, Recombinant (ECD, hFc Tag) in a functional ELISA.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence Mouse PD-1, amino acids Met1-Gln167 (Accession # NP_032824.1) with a C-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Activity Measured by its ability to bind PD-L1 Protein, Mouse, Recombinant (ECD, hFc Tag) in a functional ELISA.
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.