Learn More
ACROBiosystems Mouse PD-L1 200ug
Supplier:  ACROBiosystems PDLM82E7200UG
Encompass_Preferred
Biotinylated Mouse PD-L1 Protein HisAvitag (PDL-M82E7) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Thr 238 (Accession # Q9EP73).
Catalog No. 50-253-4309
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.