Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse PD-L1/B7-H1 Biotinylated Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. BAF1019
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1019 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1019 Supplier R&D Systems Supplier No. BAF1019

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody

PD-L1/B7-H1 Polyclonal specifically detects PD-L1/B7-H1 in Mouse samples. It is validated for Flow Cytometry∣Western Blot.

Specifications

Antigen PD-L1/B7-H1
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells
Gene Accession No. Q9EP73
Gene Alias Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1
Gene Symbols CD274
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse PD-L1/B7-H1 Phe19-Thr238 Accession # Q9EP73
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29126
Test Specificity Detects mouse PD-L1/B7-H1 in Western blots. In Western blots, approximately 15% cross-reactivity with recombinant human (rh) B7-H1 is observed and less than 1% cross-reactivity with rhB7-H3, rhB7-H2, and recombinant mouse B7-H2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.