Learn More
Invitrogen™ Mouse PD-L2/B7-DC ELISA Kit
Description
Mouse PD-L2/B7-DC quantitates mouse PD-L2/B7-DC in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse PD-L2/B7-DC.
Specifications
Specifications
| Accession Number | B7 dendritic cell molecule; B7DC; B7-DC; bA574F11.2; Btdc; Butyrophilin B7-DC; butyrophilin-like protein; CD273; F730015O22Rik; MGC124039; MGC124040; PD1 ligand 2; PD-1 ligand 2; PD-1-ligand 2; PDCD1 ligand 2; PDCD1L2; PDCD1LG2; Pdl2; PD-L2; Programmed cell death 1 ligand 2; programmed death ligand 2 |
| Assay Range | 1.9 to 400 ng/mL |
| Assay Sensitivity | 1.9 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 58205 |
| Gene Symbol | PDCD1LG2 |
| Interassay CV | <12% |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.