Learn More
ACROBiosystems ACROBiosystems Mouse PD-L2 / B7-DC Protein, His Tag
Supplier:  ACROBiosystems PD2M52E31MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse PD-L2, His Tag (PD2-M52E3) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Arg 219 (Accession # NP_067371).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.4 kDa. The protein migrates as 37-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human PD-1, Fc Tag (Cat. No. PD1-H82F1) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.2 μg/well) plate, can bind Mouse PD-L2, His Tag (Cat. No. PD2-M52E3) with a linear range of 20-156 ng/mL (QC tested).;Immobilized Mouse PD-L2, His Tag (Cat. No. PD2-M52E3) at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse PD-1, Fc,Avitag (Cat. No. PD1-M82F4) with a linear range of 0.156-2.5 μg/mL (Routinely tested).
Catalog No. 50-210-6678
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.