Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse PDGFRL Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7195268
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9739SP 25 μg
MAB9739100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9739SP Supplier R&D Systems Supplier No. MAB9739SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

PDGFRL Monoclonal specifically detects PDGFRL in Mouse samples. It is validated for Neutralization.

Specifications

Antigen PDGFRL
Applications Neutralization
Classification Monoclonal
Clone 1010639
Conjugate Unconjugated
Dilution Neutralization 4-24 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q6PE55
Gene Alias PDGFR-like protein, PDGRL, platelet-derived growth factor receptor-like, platelet-derived growth factor receptor-like protein, platelet-derived growth factor-beta-like tumor suppressor, PRLTS, PRLTSPDGF receptor beta-like tumor suppressor
Host Species Rat
Immunogen Human embryonic kidney cell HEK293-derived mouse PDGFRL, Gln22-Ser375, Accession # Q6PE55
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5157
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.