Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Mouse Pgf (P49764, 142 amino acids) Partial Recombinant Protein

Catalog No. 89956585 Shop All Abnova Corporation Products
Change view
Click to view available options
10μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
89-956-585 10μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-956-585 Supplier Abnova™ Supplier No. P4481

Used for Func, SDS-PAGE

Specifications

Accession Number P49764
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 18654
Molecular Weight (g/mol) 32.6kDa
Name Pgf (Mouse) Recombinant Protein
Preparation Method Escherichia coli expression system
Quantity 10 ug
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with sterilized 20 mM acetic acid, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg
Gene Alias AI854365/PIGF/Plgf
Common Name Pgf
Gene Symbol Pgf
Biological Activity Determined by its ability to chemoattract human monocytes of using a concentration range of 0.5-10ng/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Purity or Quality Grade ≥95% by SDS-PAGE and HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.