Learn More
Abcam Mouse PLTP ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB2138831X96TESTS
Encompass_Preferred
Mouse PLTP ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse PLTP in Mouse in Cell Lysate, Cell culture supernatant, EDTA Plasma, Heparin Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2458518
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.