Learn More
ACROBiosystems Monoclonal Anti-pre-Gc protein (SFTS virus (HN6)) Antibody, Mouse IgG1 (13E1)
Supplier:  ACROBiosystems PRNY315100UG
Encompass_Preferred
Immobilized SFTS virus pre-Gc Protein (HN6), His Tag (Cat. No. PRN-S52H3) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-pre-Gc protein (SFTS virus (HN6)) Antibody, Mouse IgG1 (13E1) (Cat. No. PRN-Y315) with a linear range of 0.1-2 ng/mL (QC tested).
Catalog No. 50-271-9047
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.