Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse Progranulin Quantikine ELISA Kit

Catalog No. MPGRN0
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MPGRN0 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MPGRN0 Supplier R&D Systems™ Supplier No. MPGRN0
Add to Cart
Add to Cart
This item is not returnable. View return policy

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Mouse Progranulin Quantikine ELISA Kit quantifies mouse Progranulin/PGRN in cell culture supernates (50 ul), tissue culture supernates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).

Specifications

Assay Range 0.63 - 20 ng/mL
Assay Sensitivity 0.071 ng/mL
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Specificity Natural and Recombinant Mouse Progranulin
For Use With (Application) ELISA
Gene ID (Entrez) 2896
Host Species Mouse
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50 uL),Tissue Culture Supernates (50 uL),Serum (10 uL),EDTA Plasma (10 uL),Heparin Plasma (10 uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym Acrogranin, CLN11, GEP, GP88, granulin precursor, GRN, PCDGF, PEPI, PGRN, Proepithelin
Target Progranulin/PGRN
Total Assay Time 4.5 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.