Learn More
Kingfisher Biotech Inc Mouse Prolactin Receptor Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1578M005
Encompass
The Mouse Prolactin Receptor yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Mouse Prolactin Receptor applications are for cell culture, ELISA standard, and Western Blot Control. Mouse Prolactin Receptor yeast-derived recombinant protein can be purchased in multiple sizes. Mouse Prolactin Receptor Specifications: (Molecular Weight: 17.4 kDa) (Amino Acid Sequence: QSPPGKPEIH KCRSPDKETF TCWWNPGSDG GLPTNYSLTY SKEGEKNTYE CPDYKTSGPN SCFFSKQYTS IWKIYIITVN ATNEMGSSTS DPLYVDVTYI VEPEPPRNLT LEVKQLKDKK TYLWVKWLPP TITDVKTGWF TMEYEIRLKS EEADEWEIHF TGHQTQFKVF DLYPGQKYLV QTRCKPDHGY WSRWGQEKSI EIPNDFTLKD (210)) (Gene ID: 19116). For research use only. Made in the USA
Catalog No. 50-253-3135
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.