Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse Prolactin Recombinant Protein, PeproTech®

Catalog No. 315161MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
315-16-1MG 1 mg
31516100UG 100 μg
315-16-10UG 10 μg
31516250UG 250 μg
31516500UG 500 μg
315-16-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 315-16-1MG Supplier Gibco™ Supplier No. 315161MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine Prolactin is a 22.5 kDa globular protein containing 198 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Prolactin (PRL) is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. PRL exerts its influence on different cell types through a signal transduction pathway which begins with the binding of the hormone to a transmembrane PRL receptor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P06879
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 19109
Molecular Weight (g/mol) 22.5 kDa
Name Mouse Prolactin
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AV290867; decidual prolactin; GHA1; growth hormone A1; Human prolactin; lactogenic prehormone (AA -30 to 199); preprolactin; pre-prolactin; Prl; Prl1a1; PRLB; PRLSD1; Prol; Prolactin; prolactin family 1, subfamily a, member 1; prolactin precursor; prolactin protein; RATPRLSD1; RNPROL
Common Name Prolactin
Gene Symbol PRL
Biological Activity Determined by its ability to induce the proliferation of rat Nb2-11 cells in the concentration range of 0.1-1.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence MLPICSAGDC QTSLRELFDR VVILSHYIHT LYTDMFIEFD KQYVQDREFM VKVINDCPTS SLATPEDKEQ ALKVPPEVLL NLILSLVQSS SDPLFQLITG VGGIQEAPEY ILSRAKEIEE QNKQLLEGVE KIISQAYPEA KGNGIYFVWS QLPSLQGVDE ESKILSLRNT IRCLRRDSHK VDNFLKVLRC QIAHQNNC
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.