Learn More
R&D Systems™ Mouse Proprotein Convertase 9/PCSK9 Quantikine ELISA Kit
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Assay Range | 62.50 to 4000 pg/mL |
| Assay Sensitivity | 21.9 pg/mL |
| Conjugate | HRP |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Cell Lysates, Cell Lysates, Plasma (EDTA, Heparin), Plasma (EDTA, Heparin), Serum, Serum |
| Specificity | Natural and Recombinant Mouse PCSK9. This kit detects 60 kDa, 53 kDa, and LDLR-complexed Recombinant Mouse PCSK9. |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 255738 |
| Gene Symbol | Pcsk9 |
| Quantity | 1 Kit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.