Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Pure-Blot anti-Goat IgG (H+L) Secondary Antibody (eB270), HRP, R&D Systems™
SDP

Catalog No. NB120525 Shop All Novus Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120525 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120525 Supplier R&D Systems Supplier No. NBP311670
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

IgG Monoclonal antibody specifically detects IgG in Goat samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen IgG
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone eB270
Conjugate HRP
Dilution Western Blot 1:000, Immunoprecipitation
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.1 mg/ml Bovine Serum Albumin (BSA, IgG and Protease free), 50% (v/v) Glycerol
Gene Alias IP Detection Reagent
Host Species Mouse
Immunogen Goat IgG
Purification Method Protein G purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Epitope Tags, Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Target Species Goat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.