Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse R-Spondin 1 Recombinant Protein, PeproTech®

Catalog No. 3153250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
315-32-50UG 50 μg
31532100UG 100 μg
315-32-1MG 1 mg
315-32-20UG 20 μg
31532250UG 250 μg
31532500UG 500 μg
315-32-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 315-32-50UG Supplier Gibco™ Supplier No. 3153250UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine R-Spondin-1 is a 27.1 kDa protein consisting of 245 amino acid residues. Due to glycosylation, R-Spondin-1 migrates at an apparent molecular weight of approximately 40.0 kDa by SDS-PAGE analysis under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene is a member of the R-spondin family and encodes a secreted activator protein with two cystein-rich, furin-like domains and one thrombospondin type 1 domain. In mice, the protein induces the rapid onset of crypt cell proliferation and increases intestinal epithelial healing, providing a protective effect against chemotherapy-induced adverse effects.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Z132
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 192199
Molecular Weight (g/mol) 27 kDa
Name Mouse R-Spondin 1
Quantity 50 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias CRISTIN3; cristin-3; cysteine-rich and single thrombospondin domain-containing protein 3; FLJ40906; hRspo1; mCristin-3; RGD1565558; Roof plate-specific spondin-1; RP11-566C13.1; RSPO; RSPO1; Rspondin; R-spondin; R-spondin 1; R-spondin homolog; R-spondin homolog (Xenopus laevis); R-spondin-1; thrombospondin type 1 domain containing gene Rspondin
Common Name RSPO1
Gene Symbol RSPO1
Biological Activity R-Spondin-1 enhances BMP-2-mediated differentiation of MC3T3-E1 cells.
Conjugate Unconjugated
Recombinant Recombinant
Sequence SRGIKGKRQR RISAEGSQAC AKGCELCSEV NGCLKCSPKL FILLERNDIR QVGVCLPSCP PGYFDARNPD MNKCIKCKIE HCEACFSHNF CTKCQEGLYL HKGRCYPACP EGSTAANSTM ECGSPAQCEM SEWSPWGPCS KKRKLCGFRK GSEERTRRVL HAPGGDHTTC SDTKETRKCT VRRTPCPEGQ KRRKGGQGRR ENANRHPARK NSKEPGSNSR RHKGQQQPQP GTTGPLTSVG PTWAQ
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.