Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse/Rat CCL5/RANTES Quantikine ELISA Kit
SDP

Catalog No. MMR00 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
2 x 96-well Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MMR00 2 x 96-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MMR00 Supplier R&D Systems™ Supplier No. MMR00
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse/Rat CCL5/RANTES Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse/Rat CCL5/RANTES Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse, rat CCL5/RANTES in cell culture supernates (50 ul), serum (25 ul), platelet-poor edta plasma (25 ul), platelet-poor citrate plasma (25 ul).

Specifications

Assay Range 7.80 - 500pg/mL
Assay Sensitivity 2pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Platelet-poor Plasma (EDTA, Citrate), Serum
Specificity Natural and Recombinant Mouse and Rat RANTES. This assay cross-reacts 1.3% with Recombinant Human RANTES.
For Use With (Application) ELISA
Gene ID (Entrez) 20304
Quantity 2 x 96-well Plates
Sample Volume Cell Culture Supernates (50uL),Serum (25uL),Platelet-poor EDTA Plasma (25uL),Platelet-poor Citrate Plasma (25uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym chemokine (C-C motif) ligand 5, D17S136Enormally T-expressed, and presumably secreted, EoCP, Eosinophil chemotactic cytokine, RANTES, SISd, SIS-delta, small inducible cytokine A5 (RANTES), small inducible cytokine subfamily A (Cys-Cys), member 5, Small-inducible cytokine A5, T cell-specific protein P228, T-cell specific protein p288, TCP228T-cell-specific protein RANTES
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.