Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse/Rat Flt-3 Ligand/FLT3L Quantikine ELISA Kit

Catalog No. MFK00
Change view
Click to view available options
Quantity:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MFK00 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MFK00 Supplier R&D Systems™ Supplier No. MFK00
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse/Rat Flt-3 Ligand Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse/Rat Flt-3 Ligand Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse, rat Flt-3 Ligand in cell culture supernates (50 ul), serum (50 ul).

Specifications

Assay Range 31.20 - 2000pg/mL
Assay Sensitivity 5pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Serum
Specificity Natural and Recombinant Mouse Flt-3 Ligand and natural Rat Flt-3 Ligand
For Use With (Application) ELISA
Gene ID (Entrez) 14256
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (50uL),Serum (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym FL, FLG3L, Flt3 ligand, Flt3L, FLT3LG, fms-related tyrosine kinase 3 ligand, SL cytokine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.