Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse/Rat IGF-I/IGF-1 Quantikine ELISA Kit
GSA_VASDP

Catalog No. MG100 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MG100 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MG100 Supplier R&D Systems™ Supplier No. MG100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse/Rat IGF-I/IGF-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse/Rat IGF-I/IGF-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse, rat IGF-I/IGF-1 in cell culture supernates (25 ul), tissue homogenates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).  Assay range: 31.2 - 2,000 pg/mL.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Assay Sensitivity 8.4 pg/mL
Content And Storage Store in refrigerator (2–8°C).
Assay Solid Phase Sandwich ELISA
Conjugate Unconjugated
Cross Reactivity < 0.5% cross-reactivity observed with available related molecules.< 50% cross-species reactivity observed with species tested.
Immunoassay Kit Format 96-well Strip Plate
Product Line R&D Systems
Product Type IGF-I Quantikine ELISA Kit
Specificity Natural and Recombinant Mouse and Rat IGF-I
Target Species Mouse
For Use With (Application) Flow Cytometry
Gene ID (Entrez) 16000
Host Species Mouse
Label or Dye Unconjugated
Quantity 1 Kit
Shipping Condition Wet Ice
Synonym IBP1, IGF1, IGF-1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, Somatomedin A, Somatomedin C, Somatomedin-C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.