Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse/Rat IL-6 ELISpot Development Module, 5 Plate
SDP

Catalog No. SEL406 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit (for 5 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SEL406 1 Kit (for 5 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SEL406 Supplier R&D Systems™ Supplier No. SEL406

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sufficient for at least five 96-well plates

Product Features:
  • An economical alternative to ELISpot Kits
  • Optimized capture and detection antibody pairings and recommended concentrations save lengthy development time
  • Generic development protocols provide direction to start an optimization protocol
  • Customize the assay to your specific needs


Module content:
  • Capture Antibody
  • Biotinylated-conjugated Detection Antibody

Specifications

Antigen IL-6
Cross Reactivity Mouse
Quantity 1 Kit (for 5 Plates)
Synonym B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Gene ID (Entrez) 3569
Gene Symbol Il6
Form Lyophilized
Product Line R&D Systems
Host Species Mouse, Rat
Product Type ELISpot Development Module
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.