Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse/Rat Inositol Monophosphatase 3/IMPAD1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7028
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7028 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7028 Supplier R&D Systems Supplier No. AF7028
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Inositol Monophosphatase 3/IMPAD1 Polyclonal specifically detects Inositol Monophosphatase 3/IMPAD1 in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Inositol Monophosphatase 3/IMPAD1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. Q80V26
Gene Alias EC 3.1.3, EC 3.1.3.25, FLJ20421, IMP 3, IMP3, IMPA3, IMPA3IMPase 3, IMPase 3, inositol monophosphatase 3, inositol monophosphatase domain containing 1, Inositol monophosphatase domain-containing protein 1, Inositol-1(or 4)-monophosphatase 3, Myo-inositol monophosphatase A3
Gene Symbols IMPAD1
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Inositol Monophosphatase 3/IMPAD1 Asn145-His356 Accession # Q80V26
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54928
Test Specificity Detects recombinant mouse Inositol Monophosphatase 3/IMPAD1 in direct ELISAs and Western blots. Detects mouse Inositol Monophosphatase 3/IMPAD1 and rat Inositol Monophosphatase 3/IMPAD1 in Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.