Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse/Rat Leptin Quantikine ELISA Kit
SDP

Catalog No. MOB00B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 Pk. (6 Plates)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MOB00B 1 Kit
SMOB00B 1 Pk. (6 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MOB00B Supplier R&D Systems™ Supplier No. MOB00B
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

solid phase ELISA designed to measure mouse or rat Leptin in cell culture supernates, tissue homogenates, serum, and plasma

Specifications

Assay Range 31.30 to 2000 pg/mL
Assay Sensitivity 5.56 pg/mL
Sample Type Cell Culture Supernatant, Mouse Plasma (EDTA), Mouse Plasma (Heparin), Mouse Serum, Rat Plasma (EDTA), Rat Plasma (Heparin), Rat Serum, Tissue Homogenates
Specificity Natural and Recombinant Mouse and Rat Leptin
Target Species Mouse, Rat
For Use With (Application) ELISA
Gene ID (Entrez) 3952
Quantity 1 Kit
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym FLJ94114, LEP, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), OB, Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS
Target Leptin/OB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.