Learn More
Description
Specifications
Specifications
| Assay Range | 31.20 - 2000pg/mL |
| Assay Sensitivity | 19.3pg/mL |
| Conjugate | HRP |
| Product Line | R&D Systems |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum |
| Specificity | Natural and Recombinant Mouse and Rat PDGF-BB. This assay cross-reacts 10.2% with Recombinant Rat PDGF-AB, 5.3% with Recombinant Human PDGF-AB, 64.7% with Recombinant Human PDGF-BB, 38.1% with natural Human PDGF and 65.2% with natural Porcine PDGF. |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 5155 |
| Quantity | 1 x 96-well Plate |
| Sample Volume | Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (25uL),Heparin Plasma (25uL) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
