Learn More
R&D Systems™ Mouse/Rat/Porcine/Canine IGF-II/IGF2 Quantikine ELISA Kit
Description
Specifications
Specifications
| Assay Range | 12.50 to 800 pg/mL |
| Assay Sensitivity | 1.87 pg/mL |
| Sample Type | Canine Serum, Mouse Serum, Porcine Serum, Rat Serum, Serum-free Cell Culture Supernatant |
| Specificity | Natural and Recombinant IGF-II. |
| Gene ID (Entrez) | 3481 |
| Host Species | Mouse, Rat, Porcine, Canine |
| Quantity | 1 Kit |
| Sample Volume | Serum-free Cell Culture Supernates (50 μL),Mouse Serum (10 μL),Rat Serum (10 μL),Canine Serum (10 μL),Porcine Serum (10 μL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Synonym | C11orf43, chromosome 11 open reading frame 43, FLJ22066, FLJ44734, GRDF, IGF2, IGF-2, IGFII, IGF-II, insulin-like growth factor 2 (somatomedin A), insulin-like growth factor II, insulin-like growth factor type 2, MSA, PEG2, PP9974, somatomedin-A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.