Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse/Rat RGM-C/Hemojuvelin Quantikine ELISA Kit
SDP

Catalog No. MRGMC0 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MRGMC0 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MRGMC0 Supplier R&D Systems™ Supplier No. MRGMC0
Add to Cart
Add to Cart
This item is not returnable. View return policy

a 4.5 hour solid phase ELISA designed to measure mouse and rat RGM-C in cell culture supernates, tissue homogenates, serum, and plasma.

The Mouse/Rat RGM-C/Hemojuvelin Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse, rat RGM-C/Hemojuvelin in cell culture supernates (50 ul), serum (13 ul), edta plasma (13 ul), heparin plasma (13 ul).

Specifications

Assay Range 31.30 - 2000pg/mL
Assay Sensitivity 4.46pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Specificity Natural and Recombinant Mouse and Rat RGM-C/Hemojuvelin
For Use With (Application) ELISA
Gene ID (Entrez) 310681
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (50uL),Serum (13uL),EDTA Plasma (13uL),Heparin Plasma (13uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym DL-M, haemojuvelin, hemochromatosis type 2 (juvenile), hemojuvelin, HFE2, HFE2AMGC23953, HJV, HJVHemochromatosis type 2 protein, JH, repulsive guidance molecule c, RGMC, RGMCRGM domain family member C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.