Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse/Rat WISP-1/CCN4 Quantikine ELISA Kit
SDP

Catalog No. MWSP10 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MWSP10 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MWSP10 Supplier R&D Systems™ Supplier No. MWSP10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse/Rat WISP-1/CCN4 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse/Rat WISP-1/CCN4 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse, rat WISP-1/CCN4 in cell culture supernates (50 ul), cell lysates (50 ul), serum (13 ul), edta plasma (13 ul), heparin plasma (13 ul), urine (50 ul).

Specifications

Assay Range 15.60 - 1000pg/mL
Assay Sensitivity 2.74pg/mL
Conjugate HRP
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and Recombinant Mouse and Rat WISP-1/CCN4
For Use With (Application) ELISA
Gene ID (Entrez) 22402
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (50uL),Cell Lysates (50uL),Serum (13uL),EDTA Plasma (13uL),Heparin Plasma (13uL),Urine (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CCN family member 4, CCN4, CCN4WISP1tc, WISP1, WISP-1, WISP1c, WISP1i, WNT1 induced secreted protein 1, Wnt-1 inducible signaling pathway protein 1, WNT1 inducible signaling pathway protein 1, wnt-1 signaling pathway protein 1, Wnt-1-induced secreted protein, WNT1-inducible-signaling pathway protein 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.