Learn More
MyBioSource Mouse ELISA Kit, Regenerating Islet-Derived Protein 3 Beta (REG3B)
SDP
Supplier:  MyBioSource MBS2000305
Encompass_Preferred
The MyBioSource Mouse Reg3b ELISA Kit is designed to detect Mouse Reg3b in cell culture supernatants, serum, plasma, tissue homogenates and other biological fluids. This sandwich ELISA kit uses a pre-coated microplate with antibodies specific for Mouse Reg3b. The kit includes a standard protein to generate a standard curve for accurate quantification. The sensitive colorimetric detection allows quantification between 31.2 pg/mL to 2000 pg/mL. This ELISA kit shows no cross-reactivity with other cytokines, and is a useful tool for detecting Mouse Reg3b for research purposes.
Catalog No. NC2876617
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.