Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse RELM alpha Recombinant Protein, PeproTech®

Catalog No. 450261MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
450-26-1MG 1 mg
45026100UG 100 μg
45026250UG 250 μg
450-26-25UG 25 μg
45026500UG 500 μg
450-26-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 450-26-1MG Supplier Gibco™ Supplier No. 450261MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine RELMalpha is a 10.0 kDa monomeric protein containing 89 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

RELM alpha, also known as Resistin-like alpha or FIZZ1, is a small cytokine (88 aminoacids, MW 9.4 kDa) produced mostly by macrophages. Its highest constitutive expression has been observed in small peritoneal macrophages, alveolar macrophages and adipose tissue macrophages. It is strongly induced by IL-4 and IL-13. Together with Arginase 1 and Ym-1, RELM alpha is considered one of the hallmarks of the M2 polarization of mouse macrophages, however, macrophages polarized with IL-10 (M2c) express neither RELM alpha nor Arginase 1. Also, the expression patterns of RELM alpha and Arginase 1 in untreated tissue macrophages significantly differ. Elevated levels of RELM alpha may indicate parasitic infection. Humans do not have RELM alpha gene but human Resistin resembles mouse RELM alpha in respect to sequence, tissue distribution and function. Other members of RELM family in mouse: RELM beta, RELM gamma and Resistin display different tissue distribution and functional properties. RELM alpha plays an important role in parasite infection, mucosal inflammation and allergy.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9EP95
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 57262
Molecular Weight (g/mol) 10 kDa
Name Mouse RELM alpha
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 1810019L16Rik; cysteine-rich secreted protein A12-gamma; Cysteine-rich secreted protein FIZZ1; Fizz1; Fizz-1; FIZZ2; found in inflammatory zone 1; Himf; HXCP2; Hypoxia-induced mitogenic factor; parasite-induced macrophage novel gene 1 protein; Pmng1; RELMa; RELMalpha; RELM-alpha; RELMb; RELMbeta; RELM-beta; resistin like alpha; resistin-like alpha; Retnla; Xcp2
Common Name RELM alpha
Gene Symbol Retnla
Conjugate Unconjugated
Recombinant Recombinant
Sequence MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.