Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse RELM beta Recombinant Protein, PeproTech®

Catalog No. 45026B25UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45026B25UG 25 μg
45026B100UG 100 μg
450-26B-1MG 1 mg
45026B250UG 250 μg
45026B500UG 500 μg
450-26B-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 45026B25UG Supplier Gibco™ Supplier No. 45026B25UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine RELMbeta is an 18.0 kDa protein, consisting of two identical 83 amino acid polypeptide chains linked by a single disulfide bond. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Adipose tissue is an energy reserve that is strictly regulated in animals. Adipose cells produce and secrete numerous physiologically important proteins, such as lipoprotein lipase (LPL), leptin, adipocyte complement related protein of 30 kDa (Acrp30), perilipin, and resistin. Resistin, also known as Adipocyte Secreted Factor (ADSF) and FIZZ3, is a cytokine that is specifically secreted by adipocytes. While the function of resistin is not well understood, it appears to have key paracrine roles and has a suggested role in type 2 diabetes. Studies show that in obese, diabetic mice the levels of serum resistin are increased, which has an antagonistic effect on insulin. Additional studies have shown that resistin causes insulin resistance and glucose intolerance in mice. Resistin gene expression requires C/EBP alpha, which is necessary and sufficient for expression. Studies show that the coactivators CBP and p300, which have been shown to display histone acetyltransferase (HAT) activity, abundantly acetylate the histones at the resistin promoter suggesting their role in activating chromatin for resistin transcription. Numerous PPAR gamma and RXR ligands have been shown to downregulate resistin expression in adipocytes.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q99P86
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 57263
Molecular Weight (g/mol) 18 kDa
Name Mouse RELM beta
Quantity 25 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 9030012B21Rik; C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich protein precursor 2; CCRG; Colon and small intestine-specific cysteine-rich protein; colon carcinoma-related gene protein; cysteine-rich secreted A12-alpha-like protein 1; cysteine-rich secreted protein A12-alpha-like 1; Cysteine-rich secreted protein A12-beta; cysteine-rich secreted protein FIZZ2; FIZZ1; Fizz2; found in inflammatory zone 1; found in inflammatory zone 2; HXCP2; Relmb; RELMbeta; RELM-beta; resistin like beta; resistin-like beta; resistin-like molecule beta; RETNL2; RETNLB; RSNB; UNQ408/PRO770; XCP2; Xcp3
Common Name RELM beta
Gene Symbol Retnlb
Conjugate Unconjugated
Recombinant Recombinant
Sequence MQCSFESLVD QRIKEALSRQ EPKTISCTSV TSSGRLASCP AGMVVTGCAC GYGCGSWDIR NGNTCHCQCS VMDWASARCC RMA
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.