Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse RELM gamma Recombinant Protein, PeproTech®

Catalog No. 45026G500UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45026G500UG 500 μg
45026G100UG 100 μg
450-26G-1MG 1 mg
45026G250UG 250 μg
45026G25UG 25 μg
450-26G-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 45026G500UG Supplier Gibco™ Supplier No. 45026G500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine RELMgamma is a 9.2 kDa monomeric protein containing 88 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

RELMgamma belongs to a unique family of tissue-specific cytokines termed FIZZ (found in inflammatory zone) and RELM. The other three known members of this family, resistin, RELMalpha, and RELMbeta, are 85-94 amino acid, secreted proteins sharing a conserved C-terminal domain, characterized by 10 cysteine residues with a unique spacing motif of C-X11-C-X8-C-X-C-X3-C-X10-C-X-C-X-C-X9-C-C. RELMgamma is most closely related to RELMalpha, but is distinctly secreted in the bone marrow, spleen, lung, and peripheral blood granulocytes. The physiological role of RELMgamma may include the promotion or regulation of promyelocytic differentiation, although the specific molecular targets of RELMgamma have not been identified.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8K426
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 245195
Molecular Weight (g/mol) 9.2 kDa
Name Mouse RELM gamma
Quantity 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Common Name RELM gamma
Gene Symbol Retnlg
Conjugate Unconjugated
Recombinant Recombinant
Sequence EGTLESIVEK KVKELLANRD DCPSTVTKTF SCTSITASGR LASCPSGMTV TGCACGYGCG SWDIRDGNTC HCQCSTMDWA TARCCQLA
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.