Learn More
Description
- Also known as: Tnfrsf19l; E430021K24Rik
- RELT is a type I transmembrane glycoprotein belonging to the TNFreceptor superfamily.
- It is primarily expressed in hematopoietic tissues and peripheral blood leukocytes.
- It has been shown to activate the NFkappaB pathway and selectively bind TNF receptor-associated factor 1 (TRAF1).
- RELT is capable of stimulating T-cell proliferation in the presence of CD3 signaling, which suggests its regulatory role in immune response.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mRelt |
| Accession Number | NP_796047 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1324 bp |
| Gene ID (Entrez) | 84957 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
