Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Renin 1 Propeptide Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7066018
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4277SP 25 μg
MAB4277 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4277SP Supplier R&D Systems Supplier No. MAB4277SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Renin Monoclonal specifically detects Renin in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Renin
Applications Western Blot
Classification Monoclonal
Clone 434717
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P06281
Gene Alias angiotensin-forming enzyme, Angiotensinogenase, EC 3.4.23, EC 3.4.23.15, FLJ10761, HNFJ2, Prorenin, REN, renin, renin precursor, renal
Gene Symbols Ren1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Renin 1 Leu22-Arg402 Accession # P06281
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5972
Test Specificity Detects mouse Renin 1 Propeptide in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with the mature form of mouse Renin 1, recombinant human Renin, recombinant mouse (rm) Cathepsin D, or rmCathepsin E is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.