Learn More
ACROBiosystems Mouse ROR1 200ug
Supplier:  ACROBiosystems RO1M82E3200UG
Encompass_Preferred
Biotinylated Mouse ROR1 Protein HisAvitag (RO1-M82E3) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # Q9Z139).
Catalog No. 50-253-4232
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.