Learn More
Sino Biological Mouse S100A8 PAb 100UL 50228-RP01-100
Supplier:  Sino Biological 50228RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse S100A8 (Catalog#50228-M07E; P27005; Pro2-Glu89). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-9513
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.