Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse S1P5/EDG-8 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB9084V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB9084V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB9084V100 Supplier R&D Systems Supplier No. FAB9084V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

S1P5/EDG-8 Monoclonal specifically detects S1P5/EDG-8 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen S1P5/EDG-8
Applications Flow Cytometry
Classification Monoclonal
Clone 1196A
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q91X56
Gene Alias Edg-8, EDG8, EDG8S1P receptor Edg-8, Endothelial differentiation G-protein-coupled receptor 8, endothelial differentiation, sphingolipid G-protein-coupled receptor, 8, LPB4, S1P receptor 5, S1P5, S1PR5, sphingosine 1-phosphate receptor 5, sphingosine 1-phosphate receptor EDG8, Sphingosine 1-phosphate receptor Edg-8, sphingosine-1-phosphate receptor 5, SPPR-1, SPPR-2
Host Species Rabbit
Immunogen Mouse S1P 5EDG-8 peptide corresponding to the N-terminal extracellular sequence Accession # Q91X56
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53637
Test Specificity Detects mouse S1P 5EDG-8 in Western blots.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.