Learn More
Kingfisher Biotech Inc Mouse SCF Recombinant Protein
Supplier: Kingfisher Biotech Inc RP1301M500
The Mouse SCF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Mouse SCF applications are for cell culture, ELISA standard, and Western Blot Control. Mouse SCF yeast-derived recombinant protein can be purchased in multiple sizes. Mouse SCF Specifications: (Molecular Weight: 18.4 kDa) (Amino Acid Sequence: KEICGNPVTD NVKDITKLVA NLPNDYMITL NYVAGMDVLP SHCWLRDMVI QLSLSLTTLL DKFSNISEGL SNYSIIDKLG KIVDDLVLCM EENAPKNIKE SPKRPETRSF TPEEFFSIFN RSIDAFKDFM VASDTSDCVL SSTLGPEKDS RVSVTKPFML PPVAA (165)) (Gene ID: 17311). For research use only. Made in the USA
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.