Learn More
Sino Biological Mouse SELPLG MAb 100TESTS 50770-R001-F-100
Supplier:  Sino Biological 50770R001F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse PSGL-1/CD162 (rM PSGL-1/CD162; Catalog#50770-MCCH; Q62170; Met1-Cys307) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-241-0379
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.