Learn More
Sino Biological Semaphorin 4D / SEMA4D / CD100 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 50642RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Semaphorin 4D / SEMA4D / CD100 (rM Semaphorin 4D / SEMA4D / CD100; Catalog#50642-M08H; NP_038688.2; Met 1-Arg 733). Semaphorin 4D / SEMA4D / CD100 specific IgG was purified by Mouse Semaphorin 4D / SEMA4D / CD100 affinity chromatography.
Catalog No. 50-255-3170
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.