Learn More
Description
- Sema3d sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3D [ Mus musculus (house mouse) ]
- Also known as: 4631426B19Rik
- Sema3D is a member of the semaphorin III family of secreted signaling proteins that are involved in axon guidance during neuronal development.
- It contains an N-terminal Sema domain, an immunoglobulin like domain and a C-terminal basic domain.
- Sema3D binds neuropilin and plays an important role in cardiovascular development.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mSema3d |
| Accession Number | NP_083158 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 2347 bp |
| Gene ID (Entrez) | 223117 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
