Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse Serpin A8/Angiotensinogen ELISA Kit (Chemiluminescence)
SDP

Catalog No. NBP266667 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266667 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266667 Supplier R&D Systems™ Supplier No. NBP266667

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Mouse Serpin A8/Angiotensinogen ELISA Kit (Chemiluminescence) measures mouse Serpin A8/Angiotensinogen in serum, plasma and other biological fluids.

Specifications

Assay Range 125.00-8000 pg/mL
Assay Sensitivity 75.00 pg/mL
Cross Reactivity This kit recognizes Mouse AGT in samples. No significant cross-reactivity or interference between Mouse AGT and analogues was observed.
Product Line Novus
Product Type ELISA Kit (Chemiluminescence)
Sample Type Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 183
Gene Symbol AGT
Host Species Mouse
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target Serpin A8/Angiotensinogen
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.