Learn More
Abcam Mouse Serum Albumin Peptide, Synthetic, Blocking, 1 mg
Description
- Protein Name: Mouse Serum Albumin
- Type: Synthetic peptide
- Applications: BL
- Purity: >70%
- Form: Liquid
- Storage Buffer: pH: 6.75; Constituents: 0.87% Sodium chloride, 0.714% HEPES, 0.0584% EDTA, 0.001% Sorbitan monolaurate, ethoxylated
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Peptide-based Substrate |
| Target Molecule | Mouse Serum Albumin |
| Shipping Condition | Blue Ice |
| Reactivity | Mouse |
| Label or Dye | Unconjugated |
| Form | Liquid |
| Buffer | 0.87% sodium chloride, 0.714% HEPES, 0.0584% EDTA, 0.001% sorbitan monolaurate, ethoxylated, pH6.75 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -20°C. |
| Formulation | pH 6.75, 0.87% Sodium chloride, 0.714% HEPES, 0.0584% EDTA, 0.001% Sorbitan monolaurate, ethoxylated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.