Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse SEZ6/BSRP-C Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF4989
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF4989 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF4989 Supplier R&D Systems Supplier No. BAF4989
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

SEZ6/BSRP-C Polyclonal specifically detects SEZ6/BSRP-C in Mouse samples. It is validated for Western Blot.

Specifications

Antigen SEZ6/BSRP-C
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q7TSK2
Gene Alias BSRPC, BSRP-C, hSEZ-6, seizure related 6 homolog (mouse)
Gene Symbols SEZ6
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SEZ6/BSRP-C Gly19-Asn902 Accession # Q7TSK2
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124925
Test Specificity Detects mouse SEZ6/BSRP-C in Western blots. In Western blots, less than 2% cross-reactivity with recombinant mouse (rm) BSRP-A and rmBSRP-B is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Store the unopened product at -20°C to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.