Learn More
Description
- Ajap1 adherens junction associated protein 1 [ Mus musculus (house mouse) ]
- Also known as: Gm573
- The Shrew-1/AJAP1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- AJAP1 is a transmembrane protein found on the basolateral surface of epithelium.
- It is presumably expressed by multiple types of epithelium and participates in cellcell adhesion.
- AJAP1 is an accompanying transmembrane molecule that interacts with Ecadherin and appears to promote the internalization of E-cadherin upon growth factor stimulation.
- This facilitates the dissolution of adherens junctions with an increase in cell motility.
- AJAP1 is also known to complex with CD147 in nonpolar, invasive cells, suggesting that AJAP1 may play a more complex role in cell migration.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mAjap1 |
| Accession Number | NP_001092769 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1252 bp |
| Gene ID (Entrez) | 55966 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
