Learn More
Description
- Also known as: Cd33l3; EG620235; SIGLEC-I
- Siglec-15 is a cell-surface receptor that binds sialylated glycoproteins.
- It is expressed on macrophages and/or dendritic cells of human spleen and lymph nodes.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | Siglec-15 |
| Accession Number | NP_001094508 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1042 bp |
| Gene ID (Entrez) | 284266 |
| Gene Alias | CD33 antigen-like 3 SIGLEC-15, CD33 molecule-like 3, CD33L3, HsT1361, sialic acid binding Ig-like lectin 15, sialic acid-binding Ig-like lectin 15, Siglec15, Siglec-15 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
