Learn More
Description
- Sirpb1b signal-regulatory protein beta 1B [ Mus musculus (house mouse) ]
- The SIRP beta 1B insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- SIRPB1b is a member of the signalregulatory-protein (SIRP) family and belongs to the immunoglobulin superfamily.
- SIRP family members are receptor-type transmembrane glycoproteins known to be involved in the negative regulation of receptor tyrosine kinase-coupled signaling processes.
- SIRP beta 1 is expressed on cells of monocyte, macrophage or dendritic lineages.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mSirpb1b |
| Accession Number | NP_001166931 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1189 bp |
| Gene ID (Entrez) | 668101 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
