Learn More
Sino Biological Human SLC3A2 / CD98 Protein (His Tag) 1mg
Supplier:  Sino Biological 12206H07H1
Encompass_Preferred
A DNA sequence encoding the human SLC3A2 isoform 1 (P08195-1) (Arg 206-Ala 630) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-222-3334
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.