Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse Sonic Hedgehog/Shh N-Terminus Quantikine ELISA Kit
SDP

Catalog No. MSHH00
Change view
Click to view available options
Quantity:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSHH00 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MSHH00 Supplier R&D Systems™ Supplier No. MSHH00

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Sonic Hedgehog/Shh N-Terminus Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse Sonic Hedgehog/Shh N-Terminus Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse Sonic Hedgehog/Shh in cell culture supernates (50 ul), serum (50 ul), heparin plasma (50 ul).

Specifications

Assay Range 7.81 - 500pg/mL
Assay Sensitivity 2.37pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (Heparin), Serum
Specificity Natural and Recombinant Mouse Shh-N. This kit also recognizes low levels of Shh-N in Rat serum.
For Use With (Application) ELISA
Gene ID (Entrez) 20423
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),Heparin Plasma (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.