Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse Sonic Hedgehog/Shh Recombinant Protein
SDP

Catalog No. NB804544 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μg
5 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB804544 25 μg
NB804518 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB804544 Supplier Novus Biologicals™ Supplier No. NBP23533925UG
Add to Cart
Add to Cart

Recombinant Protein

The ED50, as determined by inducing alkaline phosphatase production using murine C3H10T1/2 cells, is less than 300 ng/mL.

Specifications

For Use With (Application) SDS-PAGE, Bioactivity
Gene ID (Entrez) 6469
Molecular Weight (g/mol) MolecularWeight-theroretical: 19.9 kDa
Quantity 25 μg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Endotoxin Concentration Less than 1 EU/mg of the rMuSHH as determined by LAL method.
Gene Alias HHG1, HHG-1, HLP3, HPE3, MCOPCB5sonic hedgehog (Drosophila) homolog, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
Gene Symbol SHH
Reconstitution Centrifuge prior to opening. Reconstitute with sterilized PBS or aqueous buffer to a final concentration of 0.1 mg/mL
Inhibitor Properties Novus Biologicals NovActive proteins are fully active and stringently purified to provide only the safest and highest efficacy proteins available. NovActive proteins are expressed in E. coli, purified by HPLC, QC tested by SDS-PAGE and Western Blot and validated on appropriate cell lines for bioactivity. All HPLC and bioactivity data is provided for your assurance. NovActive proteins are guaranteed to be greater than 90% pure by HPLC, and contain less than 0.005 EU/ug of endotoxin.
Research Category Biologically Active Proteins, Neuroscience, Sensory Systems, Stem Cell Signaling Pathway, Stem Cells, Vision
Purity or Quality Grade SDS-PAGE
Protein Sonic Hedgehog/Shh
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.