Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Sulfatase Modifying Factor 2/SUMF2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7065646
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3454SP 25 μg
MAB3454 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3454SP Supplier R&D Systems Supplier No. MAB3454SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Sulfatase Modifying Factor 2/SUMF2 Monoclonal specifically detects Sulfatase Modifying Factor 2/SUMF2 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Sulfatase Modifying Factor 2/SUMF2
Applications Western Blot
Classification Monoclonal
Clone 382407
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8BPG6
Gene Alias C-alpha-formyglycine-generating enzyme 2, C-alpha-formylglycine-generating enzyme 2, DKFZp566I1024, DKFZp686I1024, DKFZp686L17160, DKFZp781L1035, FGE2, MGC99485, paralog of the formylglycine-generating enzyme, pFGE, sulfatase modifying factor 2, sulfatase-modifying factor 2
Gene Symbols SUMF2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Sulfatase Modifying Factor 2/SUMF2 Gln34-Leu308 Accession # Q8BPG6
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25870
Test Specificity Detects mouse Sulfatase Modifying Factor 2/SUMF2 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human SUMF2 or recombinant mouse SUMF1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.