Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse T Cell Inhibitory Receptor (Blocking) Antibody Pack, Novus Biologicals™
SDP

Catalog No. NB397809 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
4 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397809 4 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397809 Supplier R&D Systems Supplier No. NBP312531

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

nan Monoclonal Antibody

Mouse T Cell Inhibitory Receptor (Blocking) Monoclonal specifically detects Mouse T Cell Inhibitory Receptor (Blocking) in Mouse samples. It is validated for Block/Neutralize, Blockade of Receptor-ligand Interaction, CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen Mouse T Cell Inhibitory Receptor (Blocking)
Applications Blocking Assay, Blockade of Receptor-ligand Interaction, CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation See individual datasheets
Gene Alias CTLA-4 blocking, LAG-3 blocking, PD-L1 blocking, T cell inhibitor blocking, T cell inhibitor neutralization, TIGIT blocking
Immunogen See individual datasheets
Quantity 4 Vials
Regulatory Status RUO
Target Species Mouse
Content And Storage Storage is content dependent.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.